missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CytKick™ Autosampler

Product Code. 16545890
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16545890 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16545890 Supplier Invitrogen™ Supplier No. A42901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The CytKick Autosampler is for use with the Attune NxT Acoustic Focusing Cytometer.

Features of the CytKick Autosampler include:
• Reliable—intelligent probe design minimizes clogging and carryover (0.5%) and prevents damage to the instrument
• Mixing by aspiration—mixing of sample by aspiration instead of shaking ensures homogeneity of the sample and maintains cell viability
• Easy transition—no disassembly, no additional QC, no reboot required for conversion between plates and tubes
• Maximum compatibility—96- and 384-well plates (standard and deep)
• Auto-cleaning—performs automated cleaning when the instrument is shutting down

Reliable
The CytKick Autosampler was designed to minimize system down time. Automated sampling devices are often plagued by clogging or probe crashes. The Intelligent Probe, with a 200-micron diameter that is larger than most probes available in the industry, minimizes instances of clogging.

To prevent probe crashes, which may require expensive probe replacement, the CytKick Autosampler automatically performs a probe self-calibration to help ensure consistent positioning of the probe within every well. The self-calibration verifies proper function of the probe, reduces variability in probe positioning over time to prevent failures during plate runs, and helps improve reliability over the life of the system. Self-calibration of the probe also allows the autosampler to be easily moved, so it can be shared between multiple instruments.

Maximum compatibility
The CytKick Autosampler allows the use of many different plate formats, including 96-, 384-, and deep-well plates. In addition, the CytKick Autosampler initiates automated cleaning procedures when the instrument is shutting down to help ensure proper maintenance with little user interaction.

Mixing by aspiration
Unlike other plate loaders that mix samples by shaking, which may not ensure homogeneity of the sample, the CytKick Autosampler mixes samples by aspiration and stirring, enabling uniform mixing while preserving sample viability and resulting in consistent data with CVs less than 10% across the plate.

Easy to transition
To maintain system flexibility, the manual sample probe is always accessible even when the loader is present, and switching between tubes and plates is detected via software and does not require additional user intervention.

Simple to operate
The instrument design and software were developed with customer ease-of-use in mind. The CytKick Autosampler has fluidic bottles with sensors that inform the user when the fluids need to be replaced. The autosampler’s indicator lights and warning systems have been designed to ensure that a user who has walked away from the system can easily see the system status at all times.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage
  • 1 CytKick Autosampler
  • 1 install kit
  • 1 10-Pk. of 96 well plates
Quantity Each
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Autosampler

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.