missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CytKick™ Max Tube Cooling Rack

Product Code. 16636264
Change view
Click to view available options
Quantity:
1 Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16636264 1 Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16636264 Supplier Invitrogen™ Supplier No. 100076823
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The CytKick Max Tube Cooling Rack is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer

  • The CytKick Max Tube Cooling Rack is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer.
  • It allows sample uptake from 16 1.5- and 2.0-mL Eppendorf tubes without the need to remove the caps. This cooling rack maintains samples at 2-15°C for 108-120 minutes while they are being analyzed on the instrument.
  • The rack is chilled by placing it in a refrigerator overnight prior to use
  • For a non-cooling version of this rack, please see the CytKick Max Tube Rack.
TRUSTED_SUSTAINABILITY

Specifications

Product Line CytKick
Quantity 1 Rack
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Tube Cooling Rack

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.