missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CytKick™ Max Universal Cooling Adaptor

Product Code. 17188485
Change view
Click to view available options
Quantity:
1 Adapter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17188485 1 Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17188485 Supplier Invitrogen™ Supplier No. 100081492
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The CytKick Max Universal Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer

  • The CytKick Max Universal Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer.
  • This cooling block maintains samples at 2-15°C for ∼65 minutes while they are being analyzed on the instrument.
  • The cooling adaptor is brought to temperature by placing it in a refrigerator overnight or in a freezer for a minimum of two hours prior to use.
  • For a longer cooling time (95-100 minutes), please see the CytKick Max Extended Cooling Adaptor.
TRUSTED_SUSTAINABILITY

Specifications

Product Line CytKick
Quantity 1 Adapter
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Cooling Adapter

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.