missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cyto-Plex™ Carboxylated Particles
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
171.00 € - 740.00 €
Specifications
Additive | Sodium Azide Preservative, 0.05% Tween-20 |
---|---|
Content And Storage | Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer. |
Contents | Polystyrene microspheres in water |
Color | Red |
Material | Polystyrene with Copolymer Grafted Surface |
Alert:
Contains sodium azide.
Product Code | Brand | Description | Diameter (Metric) | Quantity | Level | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Diameter (Metric) | Quantity | Level | Price | Quantity & Availability | ||
11888293
|
Thermo Scientific™
FM4CR01 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 1 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11898293
|
Thermo Scientific™
FM4CR01B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 1 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11808303
|
Thermo Scientific™
FM4CR02 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 2 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11818303
|
Thermo Scientific™
FM4CR02B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 2 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11828303
|
Thermo Scientific™
FM4CR03 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 3 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11838303
|
Thermo Scientific™
FM4CR03B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 3 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11848303
|
Thermo Scientific™
FM4CR04 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 4 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858303
|
Thermo Scientific™
FM4CR04B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 4 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11868303
|
Thermo Scientific™
FM4CR05 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 5 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11878303
|
Thermo Scientific™
FM4CR05B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 5 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11888303
|
Thermo Scientific™
FM4CR06 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 6 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11898303
|
Thermo Scientific™
FM4CR06B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 6 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11808313
|
Thermo Scientific™
FM4CR07 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 7 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11818313
|
Thermo Scientific™
FM4CR07B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 7 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11828313
|
Thermo Scientific™
FM4CR08 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 8 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11838313
|
Thermo Scientific™
FM4CR08B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 8 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11848313
|
Thermo Scientific™
FM4CR09 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 9 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858313
|
Thermo Scientific™
FM4CR09B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 9 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11868313
|
Thermo Scientific™
FM4CR10 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 10 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11878313
|
Thermo Scientific™
FM4CR10B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 10 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11888313
|
Thermo Scientific™
FM4CR11 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 11 |
171.00 €
1mL |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11898313
|
Thermo Scientific™
FM4CR11B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 11 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11808323
|
Thermo Scientific™
FM4CR12 |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 1 mL | Level 12 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11818323
|
Thermo Scientific™
FM4CR12B |
Cyto-Plex Carboxylated Particles, 4µm | 4 μm | 5 mL | Level 12 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11828343
|
Thermo Scientific™
FM5CR01 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 1 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11838343
|
Thermo Scientific™
FM5CR01B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 1 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11848343
|
Thermo Scientific™
FM5CR02 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 2 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858343
|
Thermo Scientific™
FM5CR02B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 2 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11868343
|
Thermo Scientific™
FM5CR03 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 3 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11878343
|
Thermo Scientific™
FM5CR03B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 3 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11888343
|
Thermo Scientific™
FM5CR04 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 4 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11898343
|
Thermo Scientific™
FM5CR04B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 4 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11808353
|
Thermo Scientific™
FM5CR05 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 5 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11818353
|
Thermo Scientific™
FM5CR05B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 5 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11828353
|
Thermo Scientific™
FM5CR06 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 6 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11838353
|
Thermo Scientific™
FM5CR06B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 6 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11848353
|
Thermo Scientific™
FM5CR07 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 7 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858353
|
Thermo Scientific™
FM5CR07B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 7 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11868353
|
Thermo Scientific™
FM5CR08 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 8 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11878353
|
Thermo Scientific™
FM5CR08B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 8 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11888353
|
Thermo Scientific™
FM5CR09 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 9 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11898353
|
Thermo Scientific™
FM5CR09B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 9 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11808363
|
Thermo Scientific™
FM5CR10 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 10 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11818363
|
Thermo Scientific™
FM5CR10B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 10 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11828363
|
Thermo Scientific™
FM5CR11 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 11 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11838363
|
Thermo Scientific™
FM5CR11B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 11 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11848363
|
Thermo Scientific™
FM5CR12 |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 1 mL | Level 12 |
171.00 €
1mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11858363
|
Thermo Scientific™
FM5CR12B |
Cyto-Plex Carboxylated Particles, 5µm | 5 μm | 5 mL | Level 12 |
740.00 €
5mL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.- Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
- Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
- Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
- High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
- Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
- Emission can be collected in either the PE-Cy5 or APC channels
- Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
Spécification
Sodium Azide Preservative, 0.05% Tween-20 | |
Polystyrene microspheres in water | |
Polystyrene with Copolymer Grafted Surface | |
Firefli™ Fluorescent Red (635/700nm) | |
1.06g/cm³ | |
Carboxylate-Modified |
Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer. | |
Red | |
Carboxylated Particle | |
Material Safety Data Sheet (MSDS) and Package Insert Sheet | |
Product stable for a minimum of 12 months | |
≤2% CV |