missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cytochalasin A
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Spezifikation
| Quantity | 1 mg |
| Description | White |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | An anti-cytoskeletal compound. Inhibits glucose transport, actin polymerization and blocks the formation of microtubules. Inhibits cell divisiion. Inhibits HIV-1 protease |
| CAS | 14110-64-6 |
| Molecular Formula | C29H35NO5 |
| Formula Weight | 477.60 |
| Sensitivity | Light Sensitive |
| Melting Point | 191°C |
| Solubility | Soluble in DMF, DMSO, ethanol and acetone. Insoluble in water. |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur