missing translation for 'onlineSavingsMsg'
Learn More

Cytochalasin B

Product Code. 15466849
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466849 10 mg 10mg
15456849 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466849 Supplier Thermo Scientific Alfa Aesar Supplier No. J65342.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An actin-disrupting agent | CAS: 14930-96-2 | C29H37NO5 | 479.62 g/mol

Cytochalasin B is used for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts. And induces DNA fragmentation, inhibits cell division, and disrupts many cell processes. Inhibits glucose transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytochalasin B is used for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts. And induces DNA fragmentation, inhibits cell division, and disrupts many cell processes. Inhibits glucose transport.

Solubility
Freely soluble in DMF, DMSO or ethanol at room temperature

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14930-96-2
Molecular Formula C29H37NO5
Molecular Weight (g/mol) 479.62
MDL Number MFCD00077704
InChI Key GBOGMAARMMDZGR-UHFFFAOYNA-N
Synonym cytochalasin b, cytochalasin b from helminthosporium dematioideum
PubChem CID 44634701
IUPAC Name 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
SMILES CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C

Specifications

Melting Point 218°C to 221°C
Color White
Odor Odorless
Quantity 10 mg
UN Number UN1544
Beilstein 1096207
Merck Index 14,2790
Solubility Information Freely soluble in DMF, DMSO or ethanol at room temperature
Formula Weight 479.61
Physical Form Powder
Chemical Name or Material Cytochalasin B
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.