missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytochrome C, 90%, from horse heart muscle

Product Code. 10285190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100mg
1g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10285190 1g 1g
10571581 100mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285190 Supplier Thermo Scientific Chemicals Supplier No. 147530010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cytochrome C, CAS # 9007-43-6, is a component of the electron transport chain in mitochondria, making it an important electron transporter that forms an electron bridge between respirable substrates and oxygen.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Cytochrome C is a key player of the electron transport chain in mitochondria
• It plays an important role in apoptotic pathways
• It also plays a role in cellular oxidation in animals and plants

which make it an effective biological electron carrier. It plays a vital role in cellular oxidation in animals and plants.

Applications

• Cytochrome c shows peroxidase activity by oxidation of various electron donors such as 2,2-azino-bis (3-ethylbenzthiazoline-6-sulphonic acid) (ABTS), 2-keto-4-thiomethyl butyric acid and 4-aminoantipyrine

• It is assumed to be the functional complex utilized in low-level laser therapy (LLLT), which increases the metabolic activity and frees up more energy for the cells to repair the tissue

• It is used as an important mediator in apoptotic pathways

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1g
Packaging Glass bottle

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.