Learn More
BTX™ CytoGo™ Bioburden Tested Electroporation Cuvette
Product Code. 31927791
Change view
Click to view available options
Capacity (Metric):
20 to 90 μL
40 to 400 μL
80 to 800 μL
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
31927791 20 to 90 μL Pack of 50
31927789 40 to 400 μL Pack of 50
31927790 80 to 800 μL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31927791 Supplier BTX™ Supplier No. 450620
Bioburden and endotoxin tested, CytoGo electroporation cuvettes are designed to enable efficient transfection while maintaining the consistency required for GMP workflows.
With BTX Cuvettes Plus CytoGo traceable electroporation cuvettes, you can advance therapeutic research and development and production with confidence, ensuring consistency, compliance, and reproducibility at every stage.
  • Available in 1 mm, 2 mm, and 4 mm gap formats to support controlled and reproducible electroporation conditions across a broad range of cell-therapy-relevant cell types
  • Designed to support electric field strengths of up to 25 kV/cm
  • Each cuvette is sterile, single-use, and cytotoxin- and endotoxin-tested
  • A QR code on each cuvette provides traceability of every electroporation experiment
Available as a 50-pack, the Cuvettes Plus CytoGo™ cuvettes provide full traceability of raw materials, manufacturing and sterilization lots, and are tested for low cytotoxicity and endotoxin levels–supporting reliable scale-up from development through production
Specifications
Sterility Sterile
Type Electroporation Cuvette
Disposable Single-use
Capacity (Metric) 20 to 90 μL
Material Polycarbonate, Polished Aluminum
Quantity 50 Pk.
Width (Metric) Gap 1 mm
For Use With (Equipment) Electroporator
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing