missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytosine, 98+%

Product Code. 11487766
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487766 5 g 5g
11497766 25 g 25g
11407776 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487766 Supplier Thermo Scientific Chemicals Supplier No. A14731.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol

Cytosine is a nucleoside used for proteomics research. It is also used as an enzyme substrate or precursor of effector molecules such as cytosine sugars.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytosine is a nucleoside used for proteomics research. It is also used as an enzyme substrate or precursor of effector molecules such as cytosine sugars.

Solubility
Clear to very slightly hazy colorless to faint yellow solution at 50 mg/ml in 0.5 M HCL.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00006034
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
PubChem CID 597
ChEBI CHEBI:16040
SMILES NC1=CC=NC(=O)N1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 2637
Merck Index 14,2795
Solubility Information Clear to very slightly hazy colorless to faint yellow solution at 50mg/ml in 0.5 M HCL.
IUPAC Name 6-amino-1H-pyrimidin-2-one
Formula Weight 111.1
Percent Purity ≥98%
Chemical Name or Material Cytosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.