missing translation for 'onlineSavingsMsg'
Learn More

Cytosine, 99+%

Product Code. 15109213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15109213 100 g Glass bottle 100g
10134080 5 g Glass bottle 5g
10442845 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15109213 Supplier Thermo Scientific Acros Supplier No. 161761000
 Restricted Item
Estimated Shipment: 03-08-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00006034
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
PubChem CID 597
ChEBI CHEBI:16040
SMILES NC1=CC=NC(=O)N1

Specifications

Melting Point >300.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 24,314
Merck Index 15,2792
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkalines and acids,slightly soluble in ethanol,chloroform,,insoluble in ether and hydrocarbons
Spectrophotometry 98% min. (lambda max. 275nm,pH 1) molar absorptivity 10400+/-2%,on dry basis
IUPAC Name 6-amino-1,2-dihydropyrimidin-2-one
Formula Weight 111.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.