missing translation for 'onlineSavingsMsg'
Learn More

Cytosine beta-D-arabinofuranoside hydrochloride, 99%, crystalline, Thermo Scientific Chemicals

Product Code. 10173294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10173294 1 g Glass bottle 1g
10021704 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173294 Supplier Thermo Scientific Acros Supplier No. 227180010
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 69-74-9 | Molecular Formula: C9H14ClN3O5 | Molecular Weight: 279.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-74-9
Molecular Formula C9H14ClN3O5
Molecular Weight (g/mol) 279.68
MDL Number MFCD00012839
InChI Key KCURWTAZOZXKSJ-JBMRGDGGSA-N
Synonym cytarabine hydrochloride
PubChem CID 134129637
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 197.0°C to 198.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 10, 2778
Specific Rotation +127° to +133° (20°C, 589nm) ((c=0.7, H2O))
IUPAC Name 4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
Formula Weight 279.67
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Cytosine β-D-arabinofuranoside hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cytosine beta-D-arabinofuranoside hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.