missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CZC 25146
GREENER_CHOICE

Product Code. 18776519
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776519

Brand: Tocris Bioscience™ 6071/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent LRRK2 inhibitor

CZC 25146 is a potent LRRK2 inhibitor (IC50 values are 4.76 and 6.87 nM for for wild-type and G2019S mutant forms of LRRK2, respectively). Also inhibits PLK4, GAK, TNK1, CAMKK2 and PIP4K2. Attenuates mutant LRRK2-induced injury of cultured rodent and human neurons. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material CZC 25146
CAS 1191911-26-8
Synonym N-[2-[[5-Fluoro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]methanesulfonamide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C22H25FN6O4S
Formula Weight 488.54
Physical Form Solid
Color Pale-purple
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.