missing translation for 'onlineSavingsMsg'
Learn More

D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 10023390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10023390 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10023390 Supplier Thermo Scientific Acros Supplier No. 301892500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 41220-48-8
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C10H11NO2·HCl
MDL Number MFCD00800581
Quantity 250 mg
Synonym 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride, d-tetrahydroisoquinoline-3-carboxylic acid hydrochloride, 3r-1,2,3,4-tetrahydroisoquinole-3-carboxylic acid hydrochloride;, 3r-1,2,3,4-terahydroisoquinoline-3-carboxylic acid hydrochloride;, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride;, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride;
InChI Key FXHCFPUEIDRTMR-SBSPUUFOSA-N
SMILES C1C(NCC2=CC=CC=C21)C(=O)O.Cl
IUPAC Name (3R)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 213.66
PubChem CID 2733172
Formula Weight 213.66
Percent Purity 97%
Physical Form Powder
Chemical Name or Material D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.