missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(-)-2-Phenylglycine, 99%

Product Code. 11497597
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497597 500 g 500g
11477597 25 g 25g
11487597 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497597 Supplier Thermo Scientific Chemicals Supplier No. A15669.36
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 875-74-1 | C8H9NO2 | 151.165 g/mol

D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors. Used to prepare semisynthetic antibiotics amoxicillin.

Solubility
Soluble in water (0.3g/100mL) and aqueous acid.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875-74-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00008061
InChI Key ZGUNAGUHMKGQNY-SSDOTTSWSA-N
Synonym d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid
PubChem CID 70134
ChEBI CHEBI:44962
IUPAC Name (2R)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Melting Point 277°C (decomposition)
Beilstein 2208676
Merck Index 14,7291
Solubility Information Soluble in water (0.3g/100mL) and aqueous acid.
Quantity 500 g
Formula Weight 151.17
Optical Rotation −154° (c=1 in 1N HCl)
Percent Purity 99%
Chemical Name or Material D-(-)-2-Phenylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.