missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine, TRC

Product Code. 30421253
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421253 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421253 Supplier Toronto Research Chemicals Supplier No. TRCP32712550G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-Amino-2-phenylacetic Acid (D-Phenylglycine)
CAS 875-74-1
Purity Grade Notes HPLC
Molecular Formula C8 H9 N O2
Quantity 50 g
Synonym (2R)-2-Amino-2-phenylacetic Acid, D-Phenylglycine, Cefalexin Monohydrate Imp. A (EP), Cefaclor Imp. A (EP), Ampicillin Sodium Imp. L (EP), Benzeneacetic acid','α-amino-','(αR)-, Benzeneacetic acid','α-amino-','(R)-, Glycine','2-phenyl-','D- (8CI), (αR)-α-Aminobenzeneacetic acid, (-)-(R)-Phenylglycine, (-)-Phenylglycine, (2R)-2-Azaniumyl-2-phenylacetate, (2R)-Amino-2-phenylethanoic acid, (R)-(-)-2-Phenylglycine, (R)-2-Amino-2-phenylacetic acid, (R)-2-Amino-2-phenylethanoic acid, (R)-2-Phenyl-2-aminoethanoic acid, (R)-2-Phenylglycine, (R)-Phenylglycine, (R)-α-Phenylglycine, D-(-)-2-Phenylglycine, D-(-)-Aminophenylacetic acid, D-(-)-Phenylglycine, D-(-)-α-Aminophenylacetic acid, D-(-)-α-Phenylglycine, D-2-Phenylglycine, D-C-Phenylglycine, D-α-Aminophenylacetic acid, D-α-Phenylglycine
InChI Formula InChI=1S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H,10,11)/t7-/m1/s1
SMILES N[C@@H](C(=O)O)c1ccccc1
IUPAC Name (2R)-2-amino-2-phenylacetic acid
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.