missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Product Code. 12987197
Change view
Click to view available options
Volume (Metric):
12.5 μL
30 μL
100 μL
220 μL
250 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Packaging:
10 x 384 tips
20 x 384 tips
20 x 96 tips
Unit Size:
Pack of 1920
Pack of 3840
Pack of 7680
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Packaging Unit Size
12987197 100 μL Non-sterile Non-filtered 20 x 384 tips Pack of 7680
10089982 12.5 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
10794434 12.5 μL Sterile Non-filtered 10 x 384 tips Pack of 3840
10592285 30 μL Sterile Non-filtered 20 x 384 tips Pack of 7680
10502295 30 μL Sterile Filtered 20 x 384 tips Pack of 7680
10283293 100 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
11719048 100 μL Sterile Non-filtered 20 x 384 tips Pack of 7680
10203443 100 μL Sterile Filtered 20 x 384 tips Pack of 7680
10573625 100 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
11567459 100 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
11577459 100 μL Sterile Filtered 20 x 96 tips Pack of 1920
11507479 30 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10572285 30 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10532865 30 μL Sterile Filtered 20 x 96 tips Pack of 1920
10541715 30 μL Non-sterile Non-filtered 10 x 384 tips Pack of 3840
10485953 220 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10495953 250 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10101633 220 μL Sterile Filtered 20 x 96 tips Pack of 1920
10747314 30 μL Non-sterile Non-filtered 20 x 96 tips Pack of 1920
10171393 100 μL Sterile - 10 x 384 tips Pack of 3840
12917207 30 μL Sterile Non-filtered 20 x 96 tips Pack of 1920
10355073 30 μL Non-sterile Non-filtered 20 x 384 tips Pack of 7680
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Product Code. 12987197 Supplier Thermo Scientific™ Supplier No. 5326
  • ONLINE EXCLUSIVE PRICE
    1830.00 €
    1795.00 € / Pack of 7680
    Save 35.00 €
    1% Off
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.

Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.

Features:

  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
  • Highly polished molds and cores ensure low liquid retention
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile-filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination

Includes: 20 magazines per case

Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) Standard
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 20 x 384 tips
Quantity 7680 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus, Thermo Scientific Versette
Product Line Matrix
Show More Show Less
How are the D.A.R.T. tips packaged for the Versette Automated Liquid Handler?

The D.A.R.T. tips for the Versette Automated Liquid Handler are packaged as shown below:

Each case contains 4 sleeves and each sleeve in turn contains 5 tip magazines. Hence, each case has 20 tip magazines.

- For the 96-channel pipetting head, each case contains 96 x 20 = 1,920 tips

- For the 384-channel pipetting head, each case contains 384x 20 = 7,680 tips

Can I use any pipette tips with the Versette Automated Liquid Handler?

The Versette Automated Liquid Handler uses a closed-tip platform incorporating D.A.R.T. (Disposable Automation Research Tips) tips. D.A.R.T.s are in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. They help ensure accurate and precise pipetting across all channels and are designed to deliver a “no-leak” seal every time.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.