missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine-d7, TRC

Product Code. 30318327
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318327 10 mg 10mg
30311074 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318327 Supplier Toronto Research Chemicals Supplier No. TRCA48100310MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Alanine-D7
CAS 2483831-62-3
Molecular Formula C3 D7 N O2
Quantity 10 mg
Synonym D-Alanine-N,N,1,2,3,3,3-d7, D-Alanine-d7, (2R)-2-Aminopropanoic-d7 acid, D-Alanine-N,N,O,2,3,3,3-d7, D-Alanine-D7
InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1/i1D3,2D/hD3
SMILES [2H]OC(=O)[C@]([2H])(N([2H])[2H])C([2H])([2H])[2H]
IUPAC Name deuterio (2R)-2,3,3,3-tetradeuterio-2-(dideuterioamino)propanoate
Molecular Weight (g/mol) 96.1363
Formula Weight 96.0916
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.