missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine, MedChemExpress

Product Code. 16420518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16420518 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16420518 Supplier Medchem Express Supplier No. HY41700
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress D-Alanine is a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR.

D-Alanine is a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR.

Specifications

Chemical Name or Material D-Alanine
CAS 338-69-2
Color White
Purity Grade Notes Research
Molecular Formula C3H7NO2
Synonym (R)-Alanine Ba 2776 D-α-Alanine
Solubility Information H2O : 120 mg/mL (1346.95 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (11.22 mM; ultrasonic and warming and heat to 80°C)
SMILES N[C@@H](C(O)=O)C
Molecular Weight (g/mol) 89.09
Quantity 5 g
Formula Weight 89.09
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.