missing translation for 'onlineSavingsMsg'
Learn More

D-allo-Isoleucine, TRC

Product Code. 30392495
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392495 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392495 Supplier Toronto Research Chemicals Supplier No. TRCI82225850MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-allo-Isoleucine
CAS 1509-35-9
Molecular Formula C6H13NO2
Quantity 50 mg
Synonym D-Alloisoleucine, Alloisoleucine, D- (8CI), (2R,3S)-2-Amino-3-methylpentanoic acid, (2R,3S)-3-Methyl-2-aminopentanoic acid, D-(-)-Alloisoleucine, D-allo-Isoleucine, threo-D-Isoleucine
InChI Formula InChI=1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5+/m0/s1
SMILES CC[C@H](C)[C@@H](N)C(=O)O
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
Molecular Weight (g/mol) 131.17
Formula Weight 131.0946
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.