missing translation for 'onlineSavingsMsg'
Learn More

D-allo-Threonine, 99%

Product Code. 11456090
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456090 1 g 1g
15423597 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456090 Supplier Thermo Scientific Alfa Aesar Supplier No. H27050.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24830-94-2 | C4H9NO3 | 119.12 g/mol

They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are found as constituents of an increasing group of biologically active peptides. Stereocontrolled syntheses of chiral and racemic key intermediates to thienamycin from d-allo-threonine and trans-crotonic acid.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24830-94-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00004526
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-allo-threonine, d-allothreonine, h-d-allo-thr-oh, d--allo-threonine, r-allothreonine, 2r,3r-2-amino-3-hydroxybutanoic acid, 2r,3r-allothreonine, r-allo-threonine, dl-allo-threonine, allothreonine, d
PubChem CID 90624
ChEBI CHEBI:32826
IUPAC Name (2R,3R)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Specifications

Melting Point 268°C (decomposition)
Beilstein 1721644
Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 119.12
Optical Rotation −8.7° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material D-allo-Threonine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.