missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-alpha-Phenylglycine, 98%

Product Code. 10163370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163370 25 g Plastic bottle 25g
10275130 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163370 Supplier Thermo Scientific Chemicals Supplier No. 130630250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 875-74-1 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875-74-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00008061
InChI Key ZGUNAGUHMKGQNY-SSDOTTSWSA-N
Synonym d-phenylglycine, d---alpha-phenylglycine, h-d-phg-oh, d-2-phenylglycine, r-2-amino-2-phenylacetic acid, r-phenylglycine, r---2-phenylglycine, d-alpha-phenylglycine, 2r-2-amino-2-phenylacetic acid, 2r-amino phenyl acetic acid
PubChem CID 70134
ChEBI CHEBI:44962
IUPAC Name (2R)-2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Absorbance 0.03 max. (c=6, 1M HCl) at 420nm
Melting Point 302°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5CH(NH2)CO2H
Quantity 25 g
Beilstein 14, 460
Merck Index 15, 7402
Specific Rotation −151° to −161° (20°C, 589nm) (c=5, 1M HCl)
Solubility Information Solubility in water: 0.3g/100mL. Other solubilities: soluble in aq.sol. of inorg. solvents and alkalies,practically insoluble in alcohol
Formula Weight 151.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-α-Phenylglycine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.