missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-alpha-Tocopherol, 97+%

Product Code. 10127900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass Bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127900 100 mg Glass Bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10127900 Supplier Thermo Scientific Chemicals Supplier No. 421031000
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 59-02-9 | C29H50O2 | 430.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59-02-9
Melting Point 2.5°C to 3.5°C
Density 0.9500g/mL
Boiling Point 200°C to 220°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass Bottle
Molecular Formula C29H50O2
Refractive Index 1.5050 to 1.507
MDL Number MFCD00072045
Quantity 100 mg
Merck Index 15, 9653
Specific Gravity 0.95
Specific Rotation Condition +24° (25°C c=2,in isooctane)
Specific Rotation + 24
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone,chloroform,ether,,oils,fats
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
SMILES CC1=C(C(=C2CCC(OC2=C1C)(C)CCCC(C)CCCC(C)CCCC(C)C)C)O
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Molecular Weight (g/mol) 430.7
PubChem CID 14985
ChEBI CHEBI:18145
Formula Weight 430.7
Percent Purity 97+%
Physical Form Viscous Liquid
Chemical Name or Material D-α-Tocopherol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.