missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-alpha-Tocopheryl acetate, 97%

Product Code. 10341161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10341161 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10341161 Supplier Thermo Scientific Chemicals Supplier No. 421060010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-95-7 | C31H52O3 | 472.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 58-95-7
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C31H52O3
Refractive Index 1.4940 to 1.498
MDL Number MFCD00072042,MFCD00072052
Quantity 1 g
Merck Index 15, 9653
Specific Rotation Condition +3° (25°C c=2,in ethanol)
Specific Rotation + 3
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
InChI Key ZAKOWWREFLAJOT-UHFFFAOYNA-N
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
IUPAC Name 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydro-2H-1-benzopyran-6-yl acetate
Molecular Weight (g/mol) 472.75
PubChem CID 86472
ChEBI CHEBI:32321
Formula Weight 472.75
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material D-α-Tocopheryl acetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.