missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-alpha-Tocopherylquinone, 90-95%

Product Code. 10143211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143211 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10143211 Supplier Thermo Scientific Chemicals Supplier No. 350305000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7559-04-8 | C29H50O3 | 446.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7559-04-8
Color Amber to Red
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Molecular Formula C29H50O3
Refractive Index 1.4920 to 1.496
MDL Number MFCD00135657
Quantity 500 mg
Specific Rotation Condition +2.00 (25.00°C c=2,in isooctane)
Specific Rotation + 2
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
InChI Key LTVDFSLWFKLJDQ-UHFFFAOYNA-N
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)(O)CCC1=C(C)C(=O)C(C)=C(C)C1=O
IUPAC Name 2-chloropropanoyl chloride
Molecular Weight (g/mol) 446.71
PubChem CID 111019
Formula Weight 446.71
Percent Purity 90-95%
Physical Form Viscous Liquid
Chemical Name or Material D-α-Tocopherylquinone, 90-95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.