missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ D-AP5

Product Code. 30001053
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001053 100 mg 100mg
18024662 1 mg 1mg
18004362 10 mg 10mg
18004982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30001053 Supplier R&D Systems™ Supplier No. 0106/100
  • ONLINE EXCLUSIVE PRICE
    1396.54 €
    1239.00 € / 100mg
    Save 157.54 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective NMDA receptor antagonist; more active form of DL-AP5

Widely used competitive NMDA receptor antagonist. More active form of AP5. Also agonist at quisqualate-sensitized AP6 site where it is less potent than the L-isomer, L-AP5 (Cat. No.0107).

DL Mixture and L-isomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79055-68-8
Molecular Formula C5H12NO5P

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 197.13
Purity >99%
Molecular Weight (g/mol) 197.127
Chemical Name or Material D-AP5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.