missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ D-AP5

Click to view available options
Quantity:
1 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
Description
Widely used competitive NMDA receptor antagonist. More active form of AP5. Also agonist at quisqualate-sensitized AP6 site where it is less potent than the L-isomer, L-AP5 (Cat. No.0107).
DL Mixture and L-isomer also available.
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 197.13 |
| Purity | >99% |
| Molecular Weight (g/mol) | 197.127 |
| Chemical Name or Material | D-AP5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction