missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Arabinose, 99+%

Product Code. 10347580 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10347580

Brand: Thermo Scientific Chemicals 161450250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28697-53-2 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28697-53-2
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
Synonym alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
PubChem CID 71313478
IUPAC Name (4S,5R)-oxane-2,3,4,5-tetrol
SMILES OCC(O)[C@@H](O)[C@H](O)C=O

Specifications

Melting Point 152.0°C to 160.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 31, 34
Specific Rotation − 103.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glycerol and ether
Formula Weight 150.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material D(-)-Arabinose, p.a.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.