missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Arabinose, 99+%

Product Code. 10751241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751241 100 g Plastic bottle 100g
10347580 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751241 Supplier Thermo Scientific Chemicals Supplier No. 161451000
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28697-53-2 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 28697-53-2
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
Synonym alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
PubChem CID 71313478
IUPAC Name (4S,5R)-oxane-2,3,4,5-tetrol
SMILES OCC(O)[C@@H](O)[C@H](O)C=O

Spezifikation

Melting Point 152.0°C to 160.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31, 34
Specific Rotation − 103.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glycerol and ether
Formula Weight 150.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material D(-)-Arabinose, p.a.
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.