missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Arabinose, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 10323-20-3 |
| Molecular Formula | C5H10O5 |
| Quantity | 100 g |
| Synonym | d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5 |
| InChI Key | SRBFZHDQGSBBOR-UHFFFAOYNA-N |
| Molecular Weight (g/mol) | 150.13 |
| PubChem CID | 66308 |
| ChEBI | CHEBI:46983 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction