missing translation for 'onlineSavingsMsg'
Learn More

D-Aspartic acid 1-methyl ester, 98%

Product Code. 15440469
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440469 25 g 25g
15420469 1 g 1g
15430469 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440469 Supplier Thermo Scientific Alfa Aesar Supplier No. H63658.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65414-78-0 | C5H9NO4 | 147.13 g/mol

D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65414-78-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD03424227
InChI Key SWWBMHIMADRNIK-GSVOUGTGSA-N
Synonym h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid
PubChem CID 7020023
IUPAC Name (3R)-3-amino-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)C(CC(=O)O)N

Specifications

Quantity 25 g
Formula Weight 147.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Aspartic acid 1-methyl ester

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.