missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Aspartic acid, 99%

Product Code. 11404687
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404687 25 g 25g
11494677 5 g 5g
11414687 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404687 Supplier Thermo Scientific Chemicals Supplier No. B21184.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1783-96-6 | C4H7NO4 | 133.103 g/mol

D-Aspartic acid modulates melatonin synthesis in the pineal gland.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Aspartic acid modulates melatonin synthesis in the pineal gland.

Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1783-96-6
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00063081
InChI Key CKLJMWTZIZZHCS-UWTATZPHSA-N
Synonym d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
PubChem CID 83887
ChEBI CHEBI:17364
IUPAC Name (2R)-2-aminobutanedioic acid
SMILES C(C(C(=O)O)N)C(=O)O

Specifications

Melting Point >300°C (decomposition)
Beilstein 1723529
Merck Index 14,840
Solubility Information Slightly soluble in water,Soluble in 1N ammonium hydroxide (100mg/ml),and dilute HCl. Insoluble in ether,and alcohol.
Density 1.66
Quantity 25 g
Formula Weight 133.1
Optical Rotation −25° (c=1 in 1N HCl)
Percent Purity 99%
Chemical Name or Material D-Aspartic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.