missing translation for 'onlineSavingsMsg'
Learn More

D-Aspartic Acid Dimethyl Ester Hydrochloride, TRC

Product Code. 30347705
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347705 Supplier Toronto Research Chemicals Supplier No. TRCA7889755G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Aspartic Acid Dimethyl Ester Hydrochloride
CAS 69630-50-8
Molecular Formula C6H12ClNO4
Quantity 5 g
Synonym D-Aspartic Acid 1,4-Dimethyl Ester Hydrochloride, Dimethyl D-Aspartate Hydrochloride
InChI Formula InChI=1S/C6H11NO4.ClH/c1-10-5(8)3-4(7)6(9)11-2;/h4H,3,7H2,1-2H3;1H/t4-;/m1./s1
SMILES Cl.COC(=O)C[C@@H](N)C(=O)OC
IUPAC Name dimethyl (2R)-2-aminobutanedioate;hydrochloride
Molecular Weight (g/mol) 197.62
Formula Weight 197.0455
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.