missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Biotin, 98%

Product Code. 10043120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10043120 1 g Glass Bottle 1g
10053120 500 mg Glass Bottle 500mg
10308013 5 g Glass Bottle 5g
11956881 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10043120 Supplier Thermo Scientific Chemicals Supplier No. 230090010
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 231.0°C to 233.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Merck Index 15,1236
Specific Rotation + 89.00
Solubility Information Solubility in water: 0.22g/L (25°C). Other solubilities: more soluble in hot water and dil. alkali,0.8g/L alcohol,insoluble in most other common organic solvents
Formula Weight 244.31
Percent Purity 98%
Chemical Name or Material D(+)-Biotin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.