missing translation for 'onlineSavingsMsg'
Learn More

D-Biotin, BAKER™, J.T. Baker™

Product Code. 15588674
Change view
Click to view available options
Quantity:
1 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15588674 1 g Wide Mouth Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588674 Supplier Avantor J.T.Baker Supplier No. 27200
  • ONLINE EXCLUSIVE PRICE
    579.36 €
    368.63 € / 1g
    Save 210.73 €
    36% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Specifications

CAS 58-85-5
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C10H16N2O3S
MDL Number MFCD00005541
Quantity 1 g
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
Molecular Weight (g/mol) 244.31
PubChem CID 171548
ChEBI CHEBI:15956
Formula Weight 244.31
Chemical Name or Material D-Biotin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.