missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Biotin-d2 (pentanoic-2,2-d2), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Biotin-D2 (pentanoic acid-2,2-D2) |
| Molecular Formula | C10 D2 H14 N2 O3 S |
| Quantity | 0.005 g |
| Synonym | Biotin-d2, (+)-Biotin-d2, (3 AS,4 S,6 aR)-Hexahydro-2-oxo-1 H-thieno[3,4-d]imidazole-4-pentanoic-α.α-d2 acid, 5-[(3 AS,4 S,6 aR)-2-Oxo-1,3,3 A,4,6,6 A-hexahydrothieno[3,4-d]imidazol-4-yl]-2,2-dideuteriopentanoic acid, Biotin-D2 (pentanoic acid-2,2-D2), D-Biotin-d2 (pentanoic-2,2-d2) |
| InChI Formula | InChI=1 S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9 H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1/i4D2 |
| SMILES | [2 H]C([2 H])(CCC[C@@H]1 SC[C@@H]2 NC(=O)N[C@H]12)C(=O)O |
| IUPAC Name | 5-[(3 AS,4 S,6 aR)-2-oxo-1,3,3 A,4,6,6 A-hexahydrothieno[3,4-d]imidazol-4-yl]-2,2-dideuteriopentanoic acid |
| Molecular Weight (g/mol) | 246.323 |
| Formula Weight | 246.1007 g/mol |
| Percent Purity | 98 atom % D, also 29% 9-d1), min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction