missing translation for 'onlineSavingsMsg'
Learn More

D-Biotin, Fisher BioReagents

Product Code. 10746064
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10746064 1 g Amber Glass 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10746064 Supplier Fisher BioReagents Supplier No. BP2321
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
D-Biotin, which forms a stable complex with Avidin, is useful in the study of Avidin-Biotin interactions and in affinity chromatography.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Biotin
CAS 58-85-5
Melting Point 228.9°C
Color White
pH 4.5
Assay Percent Range ≥99 %
Packaging Amber Glass
Molecular Formula C10H16N2O3S
MDL Number MFCD00005541
Quantity 1 g
Merck Index 15, 1236
Specific Rotation +89 to +93° (+ or -)
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
Molecular Weight (g/mol) 244.31
PubChem CID 171548
ChEBI CHEBI:15956
Formula Weight 244.31
Vapor Pressure Negligible
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material D-Biotin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.