missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Cellobiose, 98+%

Product Code. 11431938
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431938 5 g 5g
11441938 25 g 25g
11451938 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431938 Supplier Thermo Scientific Chemicals Supplier No. A14553.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 528-50-7 | C12H22O11 | 342.30 g/mol

D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Melting Point 234°C (decomposition)
Assay Percent Range ≥98%
Quantity 5 g
Beilstein 93795
Merck Index 14,1961
Solubility Information Soluble in water (1g/8ml). Insoluble in ether and ethanol.
Optical Rotation +34° (c=10 in Water, 15h)
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
Formula Weight 342.3
Percent Purity ≥98%
Chemical Name or Material D-(+)-Cellobiose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.