missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Cellobiose, 98%

Product Code. 10297213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10297213 500 g Glass bottle 500g
10346410 5 g Glass bottle 5g
10162660 25 g Glass bottle 25g
10207603 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10297213 Supplier Thermo Scientific Chemicals Supplier No. 108465000
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 528-50-7 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Melting Point 239°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 g
Beilstein 31, 380
Merck Index 15, 1963
Specific Rotation Condition + 34.00 (20.00°C c=1,H2O,15 hrs.)
Specific Rotation + 34
Solubility Information (5% in water) Clear colorless
Formula Weight 342.29
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.