missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Cellobiose; ≥99%, MP Biomedicals™

Product Code. 11473240 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473240 100 g 100g
11443240 5 g 5g
11453240 10 g 10g
11463240 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11473240 Supplier MP Biomedicals Supplier No. 0210129880
  • ONLINE EXCLUSIVE PRICE
    531.00 €
    502.95 € / 100g
    Save 28.05 €
    5% Off
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cellobiose is a disaccharide composed of two glucose molecules; These are β-1,4-glycosidically linked. It is reducing sugar, which is readily soluble in polar solvents such as water and ethanol

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 100 g
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Formula Weight 342.3
Percent Purity ≥99%
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.