missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Cellobiose; 99%, MP Biomedicals™

Product Code. 11483000 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483000 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483000 Supplier MP Biomedicals Supplier No. 0210031910
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Major electron acceptor molecule in biological oxidations that acts as a coenzyme for hydride-transfer enzymes.

Specifications

CAS 528-50-7
Color White
Assay Percent Range 0.99
Molecular Formula C12H22O11
MDL Number MFCD00136034
Quantity 10 g
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 57370084
Formula Weight 342.3
Percent Purity 93%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.