missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Cellobiose, MP Biomedicals™

Product Code. 11483240 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483240 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483240 Supplier MP Biomedicals Supplier No. 0210129890
  • ONLINE EXCLUSIVE PRICE
    1660.00 €
    1569.75 € / 500g
    Save 90.25 €
    5% Off
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Readily soluble in polar solvents, such as water and ethanol. D-(+)-Cellobiose, MP Biomedicals is a disaccharide composed of two glucose molecules. It is a reducing sugar that is used as a cell culture media supplement. These are β-1,4-glycosidically

Specifications

CAS 528-50-7
Melting Point 229°C to 239°C
Molecular Formula C12H22O11
MDL Number MFCD00136034
Quantity 500 g
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
Solubility Information Soluble in water 4% (clear colorless solution); insoluble in alcohol and ether.
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Optical Rotation +34° ± 3° (c=4, water)
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 57370084
Formula Weight 342.3
Percent Purity ≥99%
Grade Not Available
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.