missing translation for 'onlineSavingsMsg'
Learn More

D-chiro Inositol, TRC

Product Code. 30136717
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136717 500 mg 500mg
30132686 1 g 1g
30005180 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136717 Supplier Toronto Research Chemicals Supplier No. TRCI666055500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-12-9
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym D-chiro-Inositol, Inositol, D-chiro- (8CI), (+)-chiro-Inositol, D-(+)-chiro-Inositol, D-Inositol
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES O[C@@H]1[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4-,5+,6+/m0/s1
Formula Weight 180.0634
Percent Purity >95%
Chemical Name or Material D-chiro Inositol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.