missing translation for 'onlineSavingsMsg'
Learn More

D-Cysteine hydrochloride monohydrate, 98%

Product Code. 11925721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11925721 250 mg Glass Bottle 250mg
11915721 500 mg Glass Bottle 500mg
11905721 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11925721 Supplier Thermo Scientific Acros Supplier No. 442782500
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 207121-46-8 | C3H10ClNO3S | 175.627 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Color White to Yellow
Packaging Glass Bottle
Quantity 250 mg
Formula Weight 177.77
Physical Form Crystalline Powder
Assay 98%
Chemical Name or Material D-Cysteine hydrochloride monohydrate
compliance-icons
Product Identifier
  • D-Cysteine hydrochloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: D-Cysteine hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.