missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Diisopropyl Tartrate, TRC

Product Code. 30370409
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30370409

Brand: Toronto Research Chemicals TRCD4597005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-(-)-Diisopropyl Tartrate
CAS 62961-64-2
Molecular Formula C16 H17 N3 O4 S2
Quantity 5 g
Synonym (2S,3S)-2,3-Dihydroxy-butanedioic Acid 1,4-Bis(1-methylethyl) Ester, (2S,3S)-2,3-Dihydroxy-butanedioic Acid Bis(1-methylethyl) Ester, [S-(R*,R*)]-2,3-Dihydroxy-butanedioic Acid Bis(1-methylethyl) Ester, (-)-DIPT, (2S,3S)-Diisopropyl tartrate, D-(-)-Tartaric acid diisopropyl ester, Diisopropyl (-)-D-tartrate, Diisopropyl (-)-tartrate, Diisopropyl (S,S)-tartrate, Diisopropyl D-(-)-tartrate, Diisopropyl D-tartrate
InChI Formula InChI=1S/C16H17N3O4S2/c1-4-19-9-10(2)24-16(19)17-15(21)13-14(20)11-7-5-6-8-12(11)25(22,23)18(13)3/h5-9,20H,4H2,1-3H3/b17-16+
SMILES CCN1C=C(C)S/C/1=N/C(=O)C2=C(O)c3ccccc3S(=O)(=O)N2C
IUPAC Name (NE)-N-(3-ethyl-5-methyl-1,3-thiazol-2-ylidene)-4-hydroxy-2-methyl-1,1-dioxo-1λ6,2-benzothiazine-3-carboxamide
Molecular Weight (g/mol) 379.454
Formula Weight 379.066
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.