missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine hydrochloride, 97%

Product Code. 15409167
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409167 100 mg 100mg
15419167 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409167 Supplier Thermo Scientific Alfa Aesar Supplier No. H52427.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2673-72-5 | C18H38ClNO2 | 335.96 g/mol

Inhibitor of protein kinase C and calmodulin-dependent enzymes, but may stimulate mast cells by activation of protein kinase C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2673-72-5
Molecular Formula C18H38ClNO2
Molecular Weight (g/mol) 335.96
MDL Number MFCD08436977
InChI Key YDIHJJLAPMAISR-UHFFFAOYNA-N
Synonym sphingosine hydrochloride, d-erythro-sphingosine hydrochloride, d-sphingosine hcl, trans-d-erythro-2-amino-4-octadecene-1,3-diol hydrochloride, 4-octadecene-1,3-diol,2-amino-,hydrochloride 1:1 , 2s,3r,4e, 4-octadecene-1,3-diol,2-amino-, hydrochloride 1:1 , 2s,3r,4e
PubChem CID 17998971
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol;hydrochloride
SMILES Cl.CCCCCCCCCCCCCC=CC(O)C(N)CO

Specifications

Quantity 100 mg
Formula Weight 335.96
Percent Purity 97%
Chemical Name or Material D-erythro-Sphingosine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.