missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-1,6-diphosphate dicalcium salt, 95%

Product Code. 15497849
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497849 250 g 250g
15477849 25 g 25g
15487849 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497849 Supplier Thermo Scientific Chemicals Supplier No. J66477.30
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6055-82-9 | C6H10Ca2O12P2 | 416.238 g/mol

D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

Solubility
Soluble in water

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6055-82-9
Molecular Formula C6H10Ca2O12P2
Molecular Weight (g/mol) 416.238
MDL Number MFCD00135875
InChI Key MKSZTTCFYSGQHA-UHFFFAOYSA-J
Synonym d-fructose, 1,6-bis dihydrogen phosphate , calcium salt 1:2, dicalcium 2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl phosphate, dicalcium 2,3,4-tris oxidanyl-5-oxidanylidene-6-phosphonatooxy-hexyl phosphate
PubChem CID 53398690
IUPAC Name dicalcium;(2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl) phosphate
SMILES C(C(C(C(C(=O)COP(=O)([O-])[O-])O)O)O)OP(=O)([O-])[O-].[Ca+2].[Ca+2]

Specifications

Color White
Quantity 250 g
Solubility Information Soluble in water
Formula Weight 418.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-Fructose-1,6-diphosphate dicalcium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.