missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Fructose, Certified AR for Analysis, Fisher Chemical™

Product Code. 10579760 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10579760 1 kg 1kg
10111620 100 g 100g
10488210 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10579760 Supplier Fisher Chemical Supplier No. F/1952/60
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D(-)-Fructose, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number 148910
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 103°C
Color White
pH 5 to 7
Packaging Plastic powder jar
Quantity 1 kg
Formula Weight 180.16g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.