missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Fucose, 98+%

Product Code. 10042960 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10042960

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D (+)-Fucose, 98+%, CAS # 3615-37-0, also known as aldehyde-D-fucose, is an enantiomer of an aldehyde-L-fucose found in plants. It shows several biological activities, including cellular adhesion. | CAS: 3615-37-0 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D (+)-Fucose can be found mainly in plants and shows several biological activities, including cellular adhesion

Applications

• D-Fucose is usually found in plants and some microorganisms, as a relatively simple glycoside comprising only a few sugar units

• It can be used as a sole carbon source in some microorganisms

• It may be used to study enzymes involved in its mutarotation to form L-fucose

• This compound is used as a reference compound in the analysis of carbohydrate metabolites

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3615-37-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00135603
InChI Key PNNNRSAQSRJVSB-DPYQTVNSSA-N
Synonym 2r,3s,4s,5r-2,3,4,5-tetrahydroxyhexanal, aldehydo-d-fucose, d +-fucose, galactose, 6-deoxy, d-galactose, 6-deoxy, fucose, d, +-fucose, d0o8ie, d-+-fucose, d-+-fucose;6-deoxy-d-galactose
PubChem CID 94270
ChEBI CHEBI:48203
IUPAC Name (2R,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanal
SMILES CC(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 144.0°C to 145.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 31, 76
Merck Index 15, 4307
Specific Rotation Condition + 76.00 (20.00°C c=4,H2O,NH3)
Specific Rotation + 76.00
Solubility Information (5% in water) Clear colorless to slightly yellow
Formula Weight 164.16
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Fucose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.