Learn More
D(+)-Fucose, 98+%
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D (+)-Fucose can be found mainly in plants and shows several biological activities, including cellular adhesion
Applications
• D-Fucose is usually found in plants and some microorganisms, as a relatively simple glycoside comprising only a few sugar units
• It can be used as a sole carbon source in some microorganisms
• It may be used to study enzymes involved in its mutarotation to form L-fucose
• This compound is used as a reference compound in the analysis of carbohydrate metabolites
Specifications
Specifications
| Melting Point | 144.0°C to 145.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (ELSD) (HPLC) |
| Packaging | Glass bottle |
| Quantity | 250 mg |
| Beilstein | 31, 76 |
| Merck Index | 15, 4307 |
| Specific Rotation Condition | + 76.00 (20.00°C c=4,H2O,NH3) |
| Specific Rotation | + 76.00 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.