missing translation for 'onlineSavingsMsg'
Learn More

α-D-Galactose 1-phosphate Dipotassium Salt Penthahydrate, MP Biomedicals™

Product Code. 15712272 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15712272 5 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15712272 Supplier MP Biomedicals Supplier No. 0522368705
  • ONLINE EXCLUSIVE PRICE
    4510.00 €
    4274.55 € / 1g
    Save 235.45 €
    5% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α-D-Galactose 1-phosphate Dipotassium Salt Penthahydrate, White Powder

  • α-D-Galactose 1-phosphate may be used in Leloir pathway of galactose metabolism research to study the process of UDP-Galactose (UDP-Gal) formation via UDP-glucose:α-D-galactose-1-phosphate uridylyltransferase(s) (EC 2.7.7.12).
  • It may be used to identify and characterize and study the functions of galactose-1-phosphate uridylyltransferase(s).
  • D-Galactose 1-phosphate is an important building block for the synthesis of nucleotide sugars that are substrates for glycosyltransferases.

Specifications

CAS 19046-60-7
Color White
Molecular Formula C6H11K2O9P
MDL Number MFCD00065475
Quantity 5 g
Beilstein 3922740
Synonym α-D-Galactopyranosyl phosphate dipotassium salt pentahydrate, Phos-α-D-gal, α-D-Galactopyranosyl 1-phosphate dipotassium salt
Solubility Information Soluble in 5% water (clear colorless solution).
InChI Key KCIDZIIHRGYJAE-YGFYJFDDSA-L
SMILES [K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name dipotassium (2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphate
Molecular Weight (g/mol) 336.32
Grade Not Available
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.